Learn More
Stoelting™ Micro Dissecting Forceps
Stoelting™ Micro Dissecting Forceps are micro tweezers that are ideal for micro dissection of tissue and vessels and processing tissue in electron microscopy procedures.
$102.90 - $265.00
Catalog Number | Mfr. No. | Length (English) | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (English) | Model | Price | Quantity & Availability | ||||
10-001-161
![]() |
Stoelting™
5210203 |
3.25 in. | Straight fine point |
Each for $265.00
|
|
||||
10-001-163
![]() |
Stoelting™
5210205 |
3.5 in. | Angle fine point |
Each for $251.00
|
|
||||
10-001-162
![]() |
Stoelting™
5210204 |
3.5 in. | X-action fine point |
Each for $216.50
|
|
||||
10-001-165
![]() |
Stoelting™
5210207 |
4 in. | Angle fine sharp |
Each for $216.50
|
|
||||
10-001-164
![]() |
Stoelting™
5210206 |
4 in. | Straight fine sharp |
Each for $216.50
|
|
||||
10-001-185
![]() |
Stoelting™
5210243 |
4 in. | 1x2 0.5 mm, Slight curve |
Each for $257.50
|
|
||||
10-001-187
![]() |
Stoelting™
5210245 |
4 in. | 1x2 Angled, 0.8 mm |
Each for $211.50
|
|
||||
10-001-186
![]() |
Stoelting™
5210244 |
4 in. | 1x2 Full curve |
Each for $211.50
|
|
||||
10-000-443
![]() |
Stoelting™
5210241P |
4 in. | 1x2 teeth, 0.5 mm |
Each for $165.90
|
|
||||
10-000-444
![]() |
Stoelting™
5210242P |
4 in. | 1x2 teeth, Slight curve, 0.8 mm |
Each for $140.70
|
|
||||
10-000-442
![]() |
Stoelting™
5210240P |
4 in. | 1x2 teeth, Straight, 0.8 mm |
Each for $132.30
|
|
||||
10-000-440
![]() |
Stoelting™
5210238P |
4 in. | Full curve, 0.5 mm |
Each for $165.90
|
|
||||
10-000-441
![]() |
Stoelting™
5210239P |
4 in. | Serrated angle, 0.8 mm |
Each for $133.00
|
|
||||
10-001-248
![]() |
Stoelting™
5210662 |
6 in. | Angled serrated |
Each for $102.90
|
|
||||
10-001-249
![]() |
Stoelting™
5210663 |
6 in. | Angled smooth |
Each for $114.80
|
|
||||
Description
Micro tweezers come in a wide variety of tip sizes, shapes, and materials. To choose the best tweezers, keep in mind the size of the animal and/or tissue. The type of cleaning and sterilization required may affect the material of choice. For example, if autoclaving is required and users are working under a microscope on a mouse, the thinnest-tipped forceps made of stainless steel, titanium, Dumostar, INOX, and Dumoxel are good choices.
Carbon steel instruments may not be the best choice for use where there is constant exposure to water. If not handled properly, Carbon steel is likely to rust if exposed to water for a length of time. Though not recommended, flash flame decontamination could be used on carbon steel tweezers because this material is less likely to be damaged by high heat. The extreme heat from flash flame decontamination changes the molecular structure of most material adversely. High heat softens metal and shortens the useful life of instruments. Consider another method of decontamination where lower heat levels can be applied to keep instruments useful for many more years.
Specifications
Neuroscience |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.