Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Essential Lab Hydrion™ (QK-1000) DR Quaternary Test Kit and Refill

Catalog No. p-4463131 Shop All Micro Essential Lab Products
Click to view available options
Type:
Quaternary Test Kit
Quaternary Test Kit Refill
Quantity:
5 Pack
Each

Provides a simple and reliable means to measure the concentration of quaternary sanitizers, specifically Alkyldimethylbenzyl ammonium compounds, Rocall II and Hyamine 3500. Keep refills on hand (15-420-1W) to minimize downtime.

  • Color matches within the Hi Range of 0-200-400-600-800-1000ppm (parts per million) and the Lo Range of 0-100-200-300-400ppm
  • Test paper measures concentrations between 0-1000ppm
  • Kit detects exhaustion of solutions that should be replaced as well as helping to avoid using excessive amounts of sanitizing agents
  • Refills available: 15-420-1W

Specifications

For Use With (Application) For concentration of quaternary sanitizers (alkyldimethyl benzyl ammonium compounds, Rocall II, Hyamine 3500)
Includes A high range color match chart with distinct matches at increments of 0 to 1000ppm and a low range color chart with clear colors at increments of 0 to 400ppm are included
Quantity Each
Type Quaternary Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.