Learn More
Micro Essential Lab pH Test Paper, in Vial
Catalog No. 14853165c Shop All Micro Essential Lab Products
Click to view available options
pH Range:
1 to 11
1 to 12
2 to 10
Catalog No. 14-853-165C
Supplier: Micro Essential Lab 4800
Only null left
Add to Cart
Add to Cart
Immediate, accurate pH test strips cut and packaged in vials
For classroom, laboratory and general pH testing applications.
  • General use pH strips
  • Available in normal or jumbo size strips
Specifications
pH Range 1 to 12
For Use With (Application) To provide the convenience of precut paper strips and a cost comparable to two vials of old style litmus paper making them perfect for classroom use
Includes Colorimetric chart
Quantity 500 Pack
Type Jumbo pH Test Paper Strip
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.