Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micro Particles Based on Melamine Resin, Carboxylate-Modified, FITC-marked, Analytical Standard, Size: 10 m, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
5 mL
Specifications
Specifications
Physical Form | Particles |
Concentration | 2.5% (solids) |
MDL Number | MFCD00197912 |
Synonym | Melamine-formaldehyde resin |
Quantity | 5 mL |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction