Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micro Spatula, Stainless Steel
SDPSCI_ED

Catalog No. S15924 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
21 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15924 21 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15924 Supplier United Scientific™ Supplier No. SSFT08
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economical option for everyday lab use.

  • This stainless-steel micro spatula for lab use features one flat end and one pointed end.
  • The tool measures 8 in. long.

Specifications

Quantity 1 Ea.
Blade Material Stainless Steel
Handle Material Stainless Steel
Material Stainless Steel
Handle Type No Handle
Length (Metric) 21 cm
Autoclavable Autoclavable
Shape Pointed End, Blunt End
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.