Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micro Tip Box, Empty
SDPSCI_ED

Catalog No. p-7225790
Click to view available options
Size:
2 to 200 μL
200 to 1000 μL
Length (Metric):
126 mm
143 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

United Scientific™ Micro Tip Boxes come in a pack of six empty micro tip boxes and hold up to 96 tips.

Pack of six empty micro tip boxes; each box holds up to 96 tips 2 to 200 μL or 200 to 1000 μL capacity.

  • Offered at 2 to 200 μL or 200 to 1000 μL tip capacity
  • Holes are uniformly spaced for use with a multi-channel micropipette
  • Transparent top covers included
  • Autoclavable

Specifications

Includes Empty Micro Tip Box with Cover
Product Type Pipette Tip Rack
Array 12 x 8
Autoclavable Autoclavable
Color Blue/Clear
Holds Micropipette Tips (200μL )
Material Polypropylene
Positions 96
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipette Tips
Size 2 to 200 μL
Length (Metric) 126 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.