Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basix™ Micro Tips, Universal Fit
Shop All Fisherbrand Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Refill
Description
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
- All racked sterile tips are shrink wrapped and certified free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors and endotoxin and DNA
- All nonsterile racked and bulk nonfiltered tips are certified free of RNase, DNase, and DNA
Specifications
Specifications
| Volume (Metric) | 10 μL |
| Sterility | Sterile |
| Color | Natural |
| Length (English) | 1.25 in. |
| Length (Metric) | 3.17cm |
| Clean Claims | DNase, RNase, and DNA Free |
| Format | Rack |
| Tip Style | Micro Point, Universal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction