Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICRO-TOUCH™ Nitrile Gloves

Catalog No. 17000316
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large

MICRO-TOUCH™ Nitrile Gloves feature a thin, yet durable nitrile construction to provide healthcare professionals the reliable protection and tactility they need. These gloves are approved for chemotherapy use.

  • Designed with a thin, yet durable nitrile formulation for dependable protection
  • Royal blue color provides greater contrast than standard light blue nitrile gloves
  • Approved for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Large
Quantity 100 Gloves/Box, 10 Boxes/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.