Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICRO-TOUCH™ Nitrile Gloves

Catalog No. 17000317 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-317 Medium
17-000-316 Large
17-000-319 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-000-317 Supplier Ansell Supplier No. 313029080

MICRO-TOUCH™ Nitrile Gloves feature a thin, yet durable nitrile construction to provide healthcare professionals the reliable protection and tactility they need. These gloves are approved for chemotherapy use.

  • Designed with a thin, yet durable nitrile formulation for dependable protection
  • Royal blue color provides greater contrast than standard light blue nitrile gloves
  • Approved for use with chemotherapy drugs
TRUSTED_SUSTAINABILITY

Specifications

Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Medium
Quantity 100 Gloves/Box, 10 Boxes/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.