Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate

Catalog No. p-8308930
Encompass_Preferred
Click to view available options
No. per Pack:
20 Plates
5

For use with Applied Biosystems™ ProFlex™ 3 x 32-well PCR System, (Cat. No. 4484073) and all Applied Biosystems standard 96-well thermal cyclers

  • Offers same thin-wall construction for heat transfer from thermal block to sample
  • Validated with Applied BiosystemsTM PCR thermal cyclers for optimal fit and performance
  • Easy-to-tear dual side tabs for simple separation into three, individual 32-well plates
  • Semi-skirted for easy handling and labeling
  • Certified human DNA-, DNase- and RNase-free
  • Standard 250μL working volume, with 300μL maximum well volume when used with adhesive film
  • Alphanumeric labeling on each 32-well plate for easy well identification
  • 32-well adhesive seal (MicroAmpTM 32-Well Clear Adhesive Film) available for efficient plate sealing
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance Certified DNA/RNase Free
Packaging 20 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
Volume (Metric) Well 0.3 mL
No. of Wells 32, 96
Volume (Metric) Working 0.2 mL
No. per Pack 20 Plates
Quantity 20 plates
Shipping Condition Room Temperature
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.