Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Candida albicans ATCC™ 10231™† Microbiologics™

For culture purposes and QC applications

Specifications

Biosafety Level 1
Certifications/Compliance CE, online certificate of analysis
ATCC® 10231
Format Lyophilized Pellet
For Use With (Application) Culture purpose and QC application
View More Specs

Products 4
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
23-021155 View Documents MicroBioLogicsSupplier Diversity Partner
0443P
KWIK-STIK 2/Pk. Pack of 2 for $111.30
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-209 View Documents MicroBioLogicsSupplier Diversity Partner
0443K
KWIK-STIK 6/Pk. Pack of 6 for $299.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-016-663 View Documents MicroBioLogicsSupplier Diversity Partner
0443L
Lyfo-Disk 6/Pk. Pack of 6 for $296.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-264495 View Documents MicroBioLogicsSupplier Diversity Partner
0443S
KWIK-STIK 10/Pk. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
N/A
Description

Description

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single strain of Candida albicans in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2, 6 or 10
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of microorganism
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 10231™†

Specifications

Specifications

1
10231
Culture purpose and QC application
2° to 8°C
CE, online certificate of analysis
Lyophilized Pellet
Candida albicans
Safety and Handling

Safety and Handling

ShelfLife : 6 to 24 Months

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit