Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Candida geochares ATCC™ 36852™†
Shop All Microbiologics Products

Click to view available options
Quantity:
6 Pellets/Vial
Description
LYFO DISK™
- The most economical reference stock culture option; lyophilized pellet containing a single strain of Candida geochares
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 10 pellets
- ATCC™ 36852™†
Specifications
Specifications
| Species | Candida geochares |
| ATCC® | ATCC 36852 |
| Form | Pellets |
| Product Type | Lyfo-Disk QC Control |
| Quantity | 6 Pellets/Vial |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction