Description
Epower
- Lyophilized microorganism preparations
- Biosafety Level: 1
- Pellets can be used individually or combined for a mixed microorganism population challenge
- Easily manipulated to deliver a wide variety of concentrations
- Available in concentrations ranging from 102 to 108 CFU per pellet
- ATCC™ 16404™†
Specifications
Specifications
| ATCC® | 16404 |
| Biosafety Level | 1 |
| Form | Lyophilized Pellet |
| Mean Assay Value | 1.0-9.9 + 3 CFU/pellet |
| Quantity | 10/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.