Learn More
Microbiologics™ Epower™ Staphylococcus aureus subsp. aureus ATCC™ 25923™†

For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination
$761.50 - $1008.00
Specifications
ATCC® | 25923 |
---|---|
Biosafety Level | 2 |
For Use With (Application) | For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination. |
Quantity | 10/Pk. |
Species | Staphylococcus aureus subsp. aureus |
Catalog Number | Mfr. No. | Format | Mean Assay Value | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Mean Assay Value | Price | Quantity & Availability | ||||
23-003-5009
![]() |
Microbiologics™
0360E3 |
Epower, E3 | 1.0-9.9E + 03 CFU/pellet |
Pack of 10 for $761.50
|
|
||||
12-111-287
![]() |
Microbiologics™
0360E3CRM |
Epower, E3 CRM | 1.0-9.9 + 3 CFU/pellet |
Each for $1,008.00
|
|
||||
12-111-288
![]() |
Microbiologics™
0360E4CRM |
Epower, E4 CRM | 1.0-9.9 + 4 CFU/pellet |
Each for $1,008.00
|
|
||||
Description
Epower
- Lyophilized microorganism preparations
- Biosafety Level: 2
- Pellets can be used individually or combined for a mixed microorganism population challenge
- Easily manipulated to deliver a wide variety of concentrations
- Available in concentrations ranging from 102 to 108 CFU per pellet
- ATCC™ 25923™†
Specifications
25923 | |
For performing quality control functions including microbial detection, equipment calibration, method validation, and bioburden determination. | |
Staphylococcus aureus subsp. aureus |
2 | |
10/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.