Learn More
Microbiologics™ Epower™ Staphylococcus epidermidis, ATCC™ 12228™†

Lyophilized pellets add convenience to special projects and internal proficiency plans
$806.50
Specifications
ATCC® | 12228 |
---|---|
Biosafety Level | 1 |
Form | Lyophilized Pellet |
Product Type | QC Microorganism |
For Use With (Application) | Lyophilized pellets add convenience to special projects and internal proficiency plans. |
Includes: Product warranty and peel-off certificate of assay
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | ATCC® | Species | Mean Assay Value | Price | Quantity & Availability | |||
23-003-5011
![]() |
Microbiologics™
0371E3 |
12228 | Staphylococcus epidermidis | 1.0-9.9E+03 CFU per pellet |
Pack of 10 for $806.50
|
|
|||
23-003-5041
![]() |
Microbiologics™
0371E8 |
12228 | Staphylococcus epidermidis | 1.0-9.9E+08 CFU per pellet |
Pack of 10 for $806.50
|
|
|||
Description
Order Info
Available in vials containing 10 lyophilized pellets, with a range of concentration from 102 to 109 per pellet
Specifications
12228 | |
Lyophilized Pellet | |
Lyophilized pellets add convenience to special projects and internal proficiency plans. | |
Vial | |
Staphylococcus epidermidis |
1 | |
QC Microorganism | |
Product warranty and peel-off certificate of assay | |
10/Pk. |
Safety and Handling
ShelfLife : 10 to 20 Months
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.