Learn More
Microbiologics™ Escherichia coli ATCC™ BAA-2469™†
Click to view available options
Quantity:
2/Pk.
6/Pk.
Product Type:
KWIK-STIK
Lyfo-Disk QC Control
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features single strain of Escherichia coli in lyophilized pellet, reservoir of hydrating fluid and inoculating swab
- For added convenience, peel-off identification label is included for easy documentation
- Available in pack of 2 or 6
- Reference stock culture option; lyophilized pellet containing single strain of Escherichia coli
- Easy to use; re-hydrate pellet and inoculate
- Available in re-sealable vial containing 6 pellets
- ATCC™ BAA-2469™†
- New Delhi metallo-beta-lactamase (NDM-1) positive; blaKPC negative by PCR; blaNDM positive by PCR; Carbepenem-resistant (Imipenem and Ertapenem)
Specifications
Specifications
ATCC® | ATCC BAA-2469 |
Product Type | KWIK-STIK |
Quantity | 6/Pk. |
Species | Escherichia coli |