Learn More
Description
Includes
Product warranty and a peel-off Certificate of Assay
EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.
- Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
- Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
- For added convenience, a peel-off Certificate of Assay is included for quality control documentation
- Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.
Specifications
Specifications
| ATCC® | 14028 |
| Biosafety Level | 2 |
| Format | EZ-CFU |
| Product Type | EZ-CFU |
| Includes | Product warranty and peel-off certificate of assay |
| Packaging Type | Vial |
| Species | Salmonella enterica subsp. enterica serovar typhimurium |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
