Learn More
Microbiologics™ EZ-CFU™ One-Step Microorganism Preparations
Lyophilized microorganism preparations deliver less than 100CFU per 0.1mL of hydrated suspension—no dilutions are required
Supplier: Microbiologics™ 0872Z
Includes: Product warranty and a peel-off Certificate of Assay
Description
EZ-CFU One-Step preparations are designed to provide the required microorganism challenge concentrations for tests such as Growth Promotion Tests, Microbial Limits Tests and Microbial Enumeration Tests. These ready-to-use, dependable, and cost-effective enumerated microorganism preparations allow laboratories to conveniently and confidently conquer a variety of mandated Pharmacopoeial challenges.
- Up to 19 tests per 2mL aliquot or 190 total tests per kit can be completed
- Produces a durable, non-foaming suspension that is ready to be added to the challenge material immediately following the hydration step (30 minutes)
- For added convenience, a peel-off Certificate of Assay is included for quality control documentation
- Traceable to reference cultures
Order Info
Supplied as two vials of a single assayed microorganism (10 lyophilized pellets in each vial) and 10 vials of hydrating fluid (2.0mL in each vial)
Specifications
Geobacillus stearothermophilus | |
EZ-CFU | |
Product warranty and peel-off certificate of assay |
12980 | |
EZ-CFU | |
Vial |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.