Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ EZ-PEC™ Microorganism Preparations
SDP

Catalog No. 23001923 Shop All Microbiologics Products
Change view
Click to view available options
Species:
Bacillus subtilis subsp. spizizenii
Candida albicans
Escherichia coli
Pseudomonas aeruginosa
Staphylococcus aureus
ATCC®:
10231
6538
6633™*
8739
9027
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Species ATCC®
23-001-923 Bacillus subtilis subsp. spizizenii 6633™*
23-002-628 Candida albicans 10231
23-002-640 Escherichia coli 8739
23-002-642 Staphylococcus aureus 6538
23-002-641 Pseudomonas aeruginosa 9027
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-001-923 Supplier Microbiologics™ Supplier No. 0486PEC
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Product warranty and peel-off Certificate of Assay

Add convenience to pharmaceutical and cosmetic testing labs needing to meet antimicrobial effectiveness or preservative efficacy testing

Predetermined mean assay value results in a final challenge concentration of 100,000 to 1,000,000CFU/1.0mL of product tested

  • Enumerated lyophilized microorganism preparations are designed to provide the required microorganism challenge concentrations for Antimicrobial Effectiveness and Preservative Efficacy Tests
  • Traceable to reference cultures

Order Info

Each kit contains 20 lyophilized pellets of a single enumerated strain plus ten vials of hydrating fluid (2mL each)

†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC™ cultures.

Specifications

ATCC® 6633™*
Format EZ-PEC
Product Type EZ-PEC
For Use With (Application) Add convenience to pharmaceutical and cosmetic testing labs needing to meet antimicrobial effectiveness or preservative efficacy testing.
Includes Product warranty and peel-off certificate of assay
Packaging Type Vial
Quantity 2 Vials of 10 Pellets
Species Bacillus subtilis subsp. spizizenii
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.