Learn More
Microbiologics™ Aspergillus oryzae ATCC™ 10124™†
Shop All Microbiologics Products
Click to view available options
Product Type:
Lyfo-Disk QC Control
ATCC®:
ATCC 10124
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single Aspergillus oryzae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in pack of 2
- The most economical reference stock culture option; lyophilized pellet containing a single Aspergillus oryzae strain
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing 6 pellets
- ATCC™ 10124™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Specifications
Species | Aspergillus oryzae |
ATCC® | ATCC 10124 |
Product Type | Lyfo-Disk QC Control |