Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Microbiologics™ Escherichia coli ATCC™ 51813™†
Shop All Microbiologics Products

Click to view available options
Product Type:
EZ-FPC
KWIK-STIK
Lyfo-Disk QC Control
QC Microorganism
Quantity:
10/Pk.
2/Pk.
6/Pk.
Description
KWIK-STIK™
- Clever design and accurate, repeatable results
- Each device features a single Escherichia coli strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
- For added convenience, a peel-off identification label is included for easy documentation
- Available in packs of 2 or 6
- The most economical reference stock culture option; lyophilized pellet containing a single microorganism strain of Escherichia coli
- Easy to use; simply re-hydrate the pellet and inoculate
- Available in a re-sealable vial containing pellets
Epower™
- Mean Assay Value: 1.0-9.9E+03 CFU per pellet
- 10 pellets of a single microorganism with a predetermined quantitative assay
- 10 pellets of a qualitative or quantitative microorganism manufactured as a food process control.Quantitative Preparations at a concentration of 1.0-9.9E+03 CFU per pellet
- ATCC™ 51813™†
Specifications
Specifications
ATCC® | ATCC 51813 |
Biosafety Level | 1 |
Certifications/Compliance | CE, online certificate of analysis |
Content And Storage | 2°C to 8°C |
Form | Lyophilized Pellet |
Product Type | KWIK-STIK |
For Use With (Application) | Culture purpose and QC application |
Quantity | 2/Pk. |
Species | Escherichia coli |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction