Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Haemophilus influenzae ATCC™ 49247™†
SDP

Catalog No. 23002219 Shop All Microbiologics Products
Click to view available options
Product Type:
KWIK-STIK
Lyfo-Disk QC Control
Quantity:
2/Pk.
5/Pk.
6/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single Haemophilus influenzae strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2 or 6
KWIK-STIK™ Plus
  • Five self-contained KWIK-STIK units with microorganism limited to two passages from reference culture
  • LYFO DISK™
    • The most economical reference stock culture option; lyophilized pellet containing a single Haemophilus influenzae strain
    • Easy to use; simply re-hydrate the pellet and inoculate
    • Available in a re-sealable vial containing 6 pellets
    • ATCC™ 49247™†

    †The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

    Specifications

    ATCC® 49247
    Biosafety Level 2
    Certifications/Compliance CE, certificate of assay
    Content And Storage 2°C to 8°C
    Form Lyophilized Pellet
    Media Type Kwik-stik™ Plus
    Product Type KWIK-STIK
    For Use With (Application) Culture purpose and QC application
    Includes 2 Passages
    Quantity 5/Pk.
    Species Haemophilus influenzae
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.