Learn More
Microbiologics™ Nocardia brasiliensis ATCC™ 19296™†
SDP
Catalog No. 23016867 Shop All Microbiologics Products
Click to view available options
ATCC®:
19296
ATCC 19296
Product Type:
KWIK-STIK
Lyfo-Disk QC Control
This item is not returnable. View return policy
This item is not returnable. View return policy
For culture purposes and QC applications
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single Nocardia brasiliensis strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single Nocardia brasiliensis strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
  • ATCC™ 19296™†
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Specifications
Species Nocardia brasiliensis
ATCC® ATCC 19296
Product Type Lyfo-Disk QC Control
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.