Learn More
Description
Lab Elite
- Delivered in an easy-to-use KWIK-STIK™ format
- Traceable to reference culture
- Highly characterized Listeria monocytogenes
- For use in ISO 17025 accredited labs and supported by the highest accreditations in the industry
- ATCC™ 19111™†
Specifications
Specifications
| ATCC® | 19111™* |
| Biosafety Level | 2 |
| Format | KWIK-STIK |
| For Use With (Application) | For validation and verification of processes and instruments. |
| Includes | Lyophilized microbe pellet, reservoir of hydrating fluid, inoculating swab (KWIK-STIK format) |
| Packaging Type | Single |
| Species | Listeria monocytogenes |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Microbiologics, Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
