Learn More
Microbiologics™ MicroScan™ Rapid Anaerobic Panel QC Set
Catalog No. 23001683 Shop All Fisherbrand Products
Click to view available options
:
KWIK-STIK; 2/Pk.
This item is not returnable. View return policy
Catalog No. 23-001-683
This product is either new or inactive, and cannot be ordered online.
This item is not returnable. View return policy
Includes
Clostridium perfringens ATCC™ 13124™†, Bacteroides fragilis ATCC™ 25285™†, Clostridium sordellii ATCC™ 9714™†, Finegoldia magna ATCC™ 29328™†
Microorganism sets for culture purposes and QC applications
  • Commercial sets contain the manufacturer recommended quality control strains for bacterial identification systems
  • Ordering microorganisms in sets simplifies the ordering process
  • Two convenient packaging options
KWIK-STIK™
  • Clever design and accurate, repeatable results
  • Each device features a single microorganism strain in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in pack of 2
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single microorganism strain
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 pellets
†The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. Microbiologics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.