Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propionibacterium acnes ATCC™ 11827™†, Microbiologics™
SDP

Catalog No. 23004844
Click to view available options
Quantity:
2/Pk.
5/Pk.
6/Pk.
Product Type:
KWIK-STIK
Lyfo-Disk QC Control
This item is not returnable. View return policy
This item is not returnable. View return policy

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single strain of Propionibacterium acnes in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2, 5, or 6
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Propionibacterium acne
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 11827™†

Specifications

ATCC® ATCC 11827
Biosafety Level 1
Certifications/Compliance CE, online certificate of analysis
Content And Storage 2°C to 8°C
Form Lyophilized Pellet
Product Type KWIK-STIK
For Use With (Application) Culture purpose and QC application
Quantity 5/Pk.
Species Propionibacterium acnes

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.