Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Staphylococcus aureus subsp. aureus ATCC™ 25923™†
SDP

Catalog No. 23016611 Shop All Microbiologics Products
Change view
Click to view available options
Product Type:
KWIK-STIK
KWIK-STIK MSSA Live Culture Positive Control
Lyfo-Disk QC Control
Quantity:
2/Pk.
6/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
23-016-611 Lyfo-Disk QC Control 6/Pk.
23-021104 KWIK-STIK 2/Pk.
23-013-562 KWIK-STIK MSSA Live Culture Positive Control 2/Pk.
23-016-157 KWIK-STIK 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Biosafety level 2 organisms pose a moderate risk of individual infection, but low risk of community infection.
Catalog No. 23-016-611 Supplier Microbiologics™ Supplier No. 0360L
Add to Cart
Add to Cart
Biosafety level 2 organisms pose a moderate risk of individual infection, but low risk of community infection.

For culture purposes and QC applications

KWIK-STIK™

  • Clever design and accurate, repeatable results
  • Each device features a single strain of Staphylococcus aureus subsp. aureus in a lyophilized pellet, a reservoir of hydrating fluid and inoculating swab
  • For added convenience, a peel-off identification label is included for easy documentation
  • Available in packs of 2, 6 or 10
LYFO DISK™
  • The most economical reference stock culture option; lyophilized pellet containing a single strain of Staphylococcus aureus subsp. aureus
  • Easy to use; simply re-hydrate the pellet and inoculate
  • Available in a re-sealable vial containing 6 or 10 pellets
  • ATCC™ 25923™†

Specifications

ATCC® ATCC 25923
Biosafety Level 2
Format LYFO DISK™
Product Type Lyfo-Disk QC Control
For Use With (Application) For culture purposes and QC applications.
For Use With (Equipment) Cepheid GeneXpert Systems only
Includes Peel-off identification label
Packaging Type Re-sealable Vial
Quantity 6/Pk.
Species Staphylococcus aureus subsp. aureus

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC. MicroBioLogics Inc. is licensed to use these trademarks and to sell products derived from ATCC cultures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.