Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcephalin 1/BRIT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981185
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1185 Supplier Bioss Supplier No. BS11227R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.

Specifications

Antigen Microcephalin 1/BRIT1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MCPH1
Gene Accession No. Q8NEM0
Gene Alias 5430437K10Rik; BRCT-repeat inhibitor of TERT expression 1; BRIT 1; BRIT1; D030046N04Rik; FLJ12847; MCPH1; MCT; Microcephalin; microcephalin 1; microcephaly, primary autosomal recessive 1; RGD1560642; truncated microcephalin
Gene Symbols MCPH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Microcephalin 1/BRIT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79648
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.