Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell AlphaTec™ 2000 Ts Plus Coveralls

Catalog No. 19500826 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-826 2X-Large
19-500-822 Small
19-500-823 Medium
19-500-824 Large
19-500-825 X-Large
19-500-827 3X-Large
19-500-828 4X-Large
19-500-829 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-500-826 Supplier Ansell Supplier No. WH20T9211106
Add to Cart
Add to Cart
This item is not returnable. View return policy

Breathable, microporous laminate suit protects against liquid and particulate biological hazards including radioactive particulates. Previously known as MICROCHEM™ by AlphaTec 2000 STANDARD.

  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Anti-static, tested according to EN 1149-5 and AATTC 76
  • Ultra Low Linting, reduced risk of contamination in critical areas
  • Silicone free, critical in spray painting applications
  • Elasticated hood, wrists, and ankles help to minimize the risk of linting and cross contamination
  • Free of natural rubber latex
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Serged, Taped
Sterility Non-sterile
Resistance Liquid, Particle, Bloodborne Pathogens
Collar Type Hooded
Certifications/Compliance (Europe) CE CATEGORY III (Type 4-B, Type 5-B, Type 6-B, EN 1073-2, EN 14126, DIN 32781, EN 1149-5)
Latex Free Yes
Model WH20-T-92-111
No. of Pockets None
For Use With (Application) Life Sciences, Automotive, Metal Fabrication, Food Processing, Utilities, Oil and Gas, Machinery, Chemical
Quantity 25/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.