Learn More
Description
- Protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
- Optimized fit, improving wearer comfort and safety
- Low Linting
- Free of natural rubber latex
Specifications
Specifications
| Product Type | Chemical Resistant Lab Coat |
| Size | 3X-Large |
| Gender | Unisex |
| Material | Microporous Polyethylene Laminate |
| Color | White |
| Closure Type | Zipper |
| Cuff Style | Elastic |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Resistance | Chemicals |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
