Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit

Catalog No. 19500814 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-814 Small
19-500-815 Medium
19-500-816 Large
19-500-817 X-Large
19-500-818 2X-Large
19-500-819 3X-Large
19-500-820 4X-Large
19-500-821 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-500-814 Supplier Ansell™ Supplier No. NV15S9210702
This item is not returnable. View return policy

Ansell™ MICROCHEM™ by AlphaTec™ Single-Use Chemical Suit provides excellent chemical protection from critical environments

  • Proven protection, filters 99.9% of particles >3 microns
  • Air and water vapour permeable (breathable) fabric helps reduce the risk of heat stress
  • Optimised body fit, improving wearer comfort and safety
  • Silicone free, critical in spray painting applications
  • Anti-static, tested according to EN 1149-5 and AATCC 76
  • Applications:
    • Body surface checking, marking and touching up
    • Painting or cleaning tools or robots
    • Unpacking of bulk goods
    • Loading and unloading of process equipment
    • Production line support and maintenance
    • Assembly and inspection of components
    • Asbestos related work
    • Handing of powders
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical-resistant Coveralls
Size Small
Gender Unisex
Material Anti-static SMS
Color Blue
Disposable Single-use
Ankle Style Attached Boots, Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel
Seam Type Serged
Resistance Anti-static, Particle
Collar Type Hooded
Certifications/Compliance EN1073-2:2002; AATCC76; Type 5 EN ISO 13982-1:2004 + A1:2010; Type 6 EN13034:2005 + A1:2009
Thickness 50 gsm
Quantity 1/Bag, 25/Cs.
Style Hooded and booted coverall
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.