Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Electrolysis of Aqueous Solution In Electrochemical Cells Kit
Shop All Aldon Products

Click to view available options
:
Electrolysis of Aqueous Solution in Electrochemical Cell
Description
- Accommodates up to 15 lab groups
- Complete kit provides everything needed such as syringes, vials, tubing, comboplates, electrodes, acids, solutions and more
Specifications
Specifications
| Certifications/Compliance | 49 CFR 173.4 |
| For Use With (Application) | To study electrolysis of aqueous solution in electrochemical cell |
| Includes | 5 x 25mL Sulfuric Acid, 1.0M solution, 5 x 25mL Sodium Chloride, 0.1M solution, 1 x 50g Sodium Chloride Crystals, sealing putty, comboplates, battery connectors with clips and indicator light, Plastic vials, 2-hole lids, Copper electrodes, Silicone tubing, Plastic pipette tips, microstands, syringes, filter paper |
| Class Size | 15 Lab Groups |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction