Learn More
Eisco™ Microcline Feldspar Mineral Specimen
SCI_ED
Catalog No. S24703 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24703 12/Pk. 2 to 3 cm
S26595 10/Pk. 2 to 3 cm
S26626 1/Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S24703 Supplier Eisco™ Supplier No. ESNG0021PK12
Add to Cart
Add to Cart
Microcline Feldspar Mineral Specimen for use in study of physical properties like hardness, cleavage, and crystallization temperature.
  • For use in classroom identification labs and exercises
  • Hardness of 6 on Mohs scale
  • Distinctly shows two planes of cleavage
  • Differing colors among samples offer visual demonstration of chemical variation
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Mineral
For Use With (Application) Study of physical properties like hardness/cleavage/crystallization temperature
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Microcline Feldspar