Learn More
Thermo Scientific™ Micrococcal Nuclease Solution (≥100 U/μL) PROMO
Description
This micrococcal nuclease is a stable liquid form of the enzyme derived from Staphylococcus aureus. Micrococcal nuclease (MNase) exhibits exo- and endo-5'-phosphodiesterase activities against DNA and RNA. This enzyme digests double-stranded, single-stranded, circular and linear nucleic acids. The highest activity is toward single-stranded nucleic acid substrates with preference for AT- or AU-rich regions. Enzymatic activity occurs at pH 7 to 10 and is strictly dependent on calcium for the digestion of RNA and DNA substrates. Micrococcal nuclease is suitable for removing nucleic acids from cell lysates, releasing chromatin-bound proteins and shearing chromatin for use in chromatin immunoprecipitation (ChIP) experiments.
Highlights:
- Digests nucleic acids (DNA and RNA)
- Effective for double-stranded, single-stranded, circular and linear nucleic acids
- Active in neutral to alkaline buffer conditions containing divalent calcium ions
- Supplied at ≥100 units/μL in 10 mM Tris-HCl, pH 7.5, 50mM NaCl, 1mM EDTA, 50% glycerol
Compatible with:
Thermo Scientific Subcellular Protein Fractionation Kit (Part No. 78840); Thermo Scientific Pierce ChIP Kit (Part No. 26156)
Recommended for:
Enzymatic DNA shearing for chromatin immunoprecipitation assays (ChIP); Removal of unwanted genomic or organellar nucleic acids from cell lysates; Degradation of nucleic acids in subcellular fractionation procedures
Specifications
Specifications
| Product Type | Cell Lysis Enzyme |
| Content And Storage | Upon receipt store at -20°C. Product is shipped with dry ice. |
| Form | Liquid |
| Reagent Type | Enzyme for Cell Lysis |
| Quantity | 150 μL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.