Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microcrystalline Cellulose, Partially Depolymerized, FCC, EP, BP, Spectrum™ Chemical
SDP

Catalog No. 18604601 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-601 100 g Poly Bottle
18-604-604 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-604-601 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CE112100GM

CE112, 9004-34-6

Spectrum™ Chemical Microcrystalline Cellulose, Partially Depolymerized, FCC, EP, BP is an excipient in pharmaceutical formulations acting as a binder in tablet formation. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-34-6
Molecular Formula C14H26O11
Molecular Weight (g/mol) 370.35
InChI Key PTHCMJGKKRQCBF-UHFFFAOYNA-N
IUPAC Name 2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-methoxyoxan-3-yl]oxy}-6-(hydroxymethyl)-5-methoxyoxane-3,4-diol
SMILES COC1OC(CO)C(OC2OC(CO)C(OC)C(O)C2O)C(O)C1O

Specifications

CAS Max % 100%
Quantity 100 g
Grade Multi-Compendial
Ignition Residue 0.1%
Loss on Drying 7%
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.