Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ ApexPro™ LC100 AP12 Nitrile Gloves

Catalog No. 19500971 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-971 X-Small
19-500-972 Small
19-500-973 Medium
19-500-974 Large
19-500-975 X-Large
19-500-976 2X-Large
19-500-977 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-500-971 Supplier Ansell Supplier No. AP121X
This item is not returnable. View return policy

Dual-layer nitrile exam gloves with extended cuff provides protection against opioids and other hazardous substances.

  • Tested against fentanyl, and gastric acid (vomit) to simulate hazardous overdose situations
  • Gloves consist of a dual layer, bi-color design (white exterior, black interior)
  • Delivers advanced barrier protection (AQL 0.65)
  • True pH balanced interior coating for improved hand health
  • Triple washed
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D6319, ASTM D6978, FDA21 CFR 177-2600, NFPA 1999:2018, ISO 12485, ISO 9001
Chemotherapeutic Rated Yes
Applications Healthcare
AQL 0.65
Color White, Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 8.3 mil
Finger Thickness (Metric) 0.21 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.9 mil
Palm Thickness (Metric) 0.15 mm
Size X-Small
Thickness (English) 8.3 mil
Thickness (Metric) 0.21 mm
FDA 510 (k) No
Model AP12-1X
Series ApexPro™ LC100 AP12
Tensile Strength 20 MPa
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.