Learn More
Description
- Black exterior and gray interior to make potentially hazardous powders more visible
- Tested against both fentanyl and gastric acid
- Dual layer, dual color design
- Advanced barrier protection (AQL 0.65)
- pH balanced interior coating for improved hand health
- Recommended for a variety of industries including:
- Chemical
- Correctional Facilities
- Emergency Medical Services
- Healthcare
- Law Enforcement
- Life Sciences
- Machinery and Equipment
- Maintenance
- Recycling and Waste Disposal
- Transportation
Specifications
Specifications
| Certifications/Compliance | ASTM D6319; FDA21 CFR 177-2600; ISO 13485; ISO 9001; NFPA 1999:2018 |
| Chemotherapeutic Rated | No |
| Chlorinated | No |
| Applications | Healthcare, Industrial, Laboratory |
| AQL | 0.65 |
| Color | Black, Gray |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 8.3 mil |
| Finger Thickness (Metric) | 0.21 mm |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
