Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell MICROFLEX™ ApexPro™ XP100 APXP12 Nitrile Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Black exterior and gray interior to make potentially hazardous powders more visible
- Tested against both fentanyl and gastric acid
- Dual layer, dual color design
- Advanced barrier protection (AQL 0.65)
- pH balanced interior coating for improved hand health
- Recommended for a variety of industries including:
- Chemical
- Correctional Facilities
- Emergency Medical Services
- Healthcare
- Law Enforcement
- Life Sciences
- Machinery and Equipment
- Maintenance
- Recycling and Waste Disposal
- Transportation
Specifications
Certifications/Compliance | ASTM D6319; FDA21 CFR 177-2600; ISO 13485; ISO 9001; NFPA 1999:2018 |
Chemotherapeutic Rated | No |
Applications | Healthcare, Industrial, Laboratory |
AQL | 0.65 |
Color | Black, Gray |
Disposable | Single-use |
Exterior Finish | Fully Textured |
Finger Thickness (English) | 8.3 mil |
Finger Thickness (Metric) | 0.21 mm |
Latex Free | Yes |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction