Learn More
Description
- Thick, 15 mil thick at the finger to resist tears and punctures
- Tested against both fentanyl and gastric acid (vomit) to simulate hazardous, real world overdose situations
- Tested against 8 commonly used chemotherapy drugs in the Central Supply/SPD Department
- Provide chemical splash protection, including chemotherapy drugs
- Premium latex provides increased range of motion and ultimate comfort
Specifications
Specifications
| Certifications/Compliance | ASTM D3578, ASTM D6978, ISO 13485, ISO 9001 |
| Chemotherapeutic Rated | Yes |
| Applications | Healthcare, Laboratory |
| AQL | 1.5 |
| Color | Blue, Green |
| Disposable | Single-use |
| Exterior Finish | Fully Textured |
| Finger Thickness (English) | 15 mil |
| Finger Thickness (Metric) | 0.38 mm |
| Laboratory/Cleanroom Use | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.