Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ DigitSafety HaloKote™ DSK

Catalog No. 19500947 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-500-947 Large
19-500-945 Small
19-500-946 Medium
19-500-948 X-Large
19-500-949 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-500-947 Supplier Ansell Supplier No. DSK3
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Thick, latex exam gloves with extended cuff are ideal for unpredictable, high risk situations.

  • Thick, 15 mil thick at the finger to resist tears and punctures
  • Tested against both fentanyl and gastric acid (vomit) to simulate  hazardous, real world overdose situations
  • Tested against 8 commonly used chemotherapy drugs in the Central Supply/SPD Department
  • Provide chemical splash protection, including chemotherapy drugs
  • Premium latex provides increased range of motion and ultimate comfort
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D3578, ASTM D6978, ISO 13485, ISO 9001
Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 1.5
Color Blue, Green
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 15 mil
Finger Thickness (Metric) 0.38 mm
Laboratory/Cleanroom Use No
Latex Free No
Material Latex
Palm Thickness (English) 13 mil
Palm Thickness (Metric) 0.33 mm
Size Large
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
FDA 510 (k) No
Model DSK-3
Series DigitSafety HaloKote™ DSK
Tensile Strength 32 MPa
Length (English) 11.8 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.