Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ UltraSense™ EC USE-880 Nitrile Gloves

Catalog No. 19177510 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-177-510 3X-Large
19-167-295F X-Small
19-167-295A Small
19-167-295B Medium
19-167-295D X-Large
19-167-295C Large
19-167-295E 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-177-510 Supplier Ansell Supplier No. USE8803XL
This item is not returnable. View return policy

Extended cuff offers extra protection over the wrist and forearm for use in high-risk environments.

High-quality nitrile offers reliable barrier protection against many hazards and infectious substances.

  • Patented ultra-soft nitrile provides the fit and feel of latex
  • Textured fingers ensure a secure grip
  • Extended, beaded cuff offers extra protection over wrist and forearm
  • Nonsterile, ambidextrous, single-use
  • Length: 11.4 in. (29 cm)
  • Fingertip thickness: 4.3 mil

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM D 6319
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chemotherapeutic Rated No
Chlorinated Yes
Applications Healthcare, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.9 mil
Palm Thickness (Metric) 0.10 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 3X-Large
Sterility Non-sterile
Thickness (English) 5.1 mil
Thickness (Metric) 0.13 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Model USE-880-3XL
Quantity 1000/Cs.
Series UltraSense™ EC USE-880
Tensile Strength 33 MPa
Length (English) 11.4 in.
Length (Metric) 29 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.