Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microglobulin beta2 Mouse anti-Human, Clone: BBM.1, MilliporeSigma™

Catalog No. MABF196825 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1968-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1968-25 Supplier MilliporeSigma Supplier No. MABF196825UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Microglobulin beta2, clone BBM.1 Antibody Inhibition, and Flow Cytometry, for the detection of Microglobulin beta2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Microglobulin beta2
Applications Western Blot, Immunohistochemistry (Paraffin), Inhibition Assays, Flow Cytometry
Classification Monoclonal
Clone BBM.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols B2M,CDABP0092,HDCMA22P
Host Species Mouse
Immunogen MOLT-4, a Human acute lymphoblastic leukemia cell line.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone BBM.1 detects Beta-2-microglobulin in human.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.