Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ MicroGuard CE™ Clean Processed Microporous Coveralls

Catalog No. 19472829 Shop All International Enviroguard Systems Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Packaging:
Bulk
Individually Packaged
This item is not returnable. View return policy
This item is not returnable. View return policy

Clean processed microporous coveralls offer the perfect balance of safety and comfort. Ideal for non-hazardous work environments where reliable liquid and particulate protection is needed.

These clean processed coveralls offer the perfect balance of safety and comfort. Microporous fabric keeps dust and liquids out while allowing air to flow freely, so you stay cool as you put in a hard day's work. These coveralls are ideal for non-hazardous work environments where you need reliable liquid and particulate protection. Coveralls are designed with tunnelized elastic to offer a more secure closure, stronger seams, and the elimination of raw edges that may generate unwanted particulates. Each coverall is individually packaged to help prevent cross-contamination.
  • Roomy, seamless shoulder area resists tears and provides more protection in key exposure area.
  • Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric.
  • Elastic back stretches with user for more comfort and better tear resistance.
  • Secure elastic wrists and ankles help keep contaminants out.
  • Environmentally friendly - can be recycled after use.

Specifications

Product Type Cleanroom Coveralls
Size X-Large
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Abrasion, Caustics, Liquid, Particle, Water
Back Style Elastic
Model MicroGuard CE™
No. of Pockets None
For Use With (Application) Liquid barrier and particulate barrier in controlled environments
Packaging Individually Packaged
Cleanroom Class Class 100 (ISO 5)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.