Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ MicroGuard MP 17 in. Microporous Boot Cover with Non-skid Sole and Elastic Closure
Click to view available options
Size:
Large
X-Large
Description
- Boot covers are designed with a non-skid sole to help reduce the risk of slips or falls
- A tear-resistant liquid and particulate barrier helps safeguard from blood penetration, various industrial chemicals, along with dust, dirt, and grime
- Breathable fabric allows moisture to escape, so your feet stay more cool and comfortable even as you sweat
- 17 in. length helps protect more of your lower legs from exposure to on-the-job contaminants
- Secure elastic closure helps keep boot covers in place throughout your workday
- Environmentally friendly; can be recycled after use
- Strong, elastic top for a secure fit
- Made of lightweight, tear-resistant polypropylene
- Passes ASTM F1670 testing for blood penetration
Specifications
Specifications
Size | Large |
Material | Multi-layer Barrier Laminate Fabric, Spunbond Polypropylene |
Latex Free | Yes |
Color | White |
Disposable | Single-use |
Sterility | Non-sterile |
Closure Type | Elastic Top with Ankle Ties |
Sole Type | Non-skid |
Seam Type | Serged |
Resistance | Static, Liquid, Grime, Dry Particulates |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction