Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        DWK Life Sciences MicroLiter 20 mm Pressure Release Crimp Top Headspace Vial Seals
    
    
    
        
            Shop All DWK Life Sciences Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Septum:
                            
                        
                        
                            
                                Aluminum, Silicone
                            
                        
                            
                                None
                            
                        
                            
                                PTFE/Butyl
                            
                        
                            
                                Tan PTFE, White Silicone
                            
                        
                    Description
Pressure release seal
                        
                        
                        
                        
                        
                        
                        
                        
                        
                    
                
                
                    
                
                
    
    
        
            
            Specifications
Specifications
| Color | Silver | 
| Material | Aluminum | 
| Septum | None | 
| No. per Pack | 100 | 
| Product Line | MicroLiter | 
| Type | Crimp Cap | 
| Closure Size | 20 mm | 
| For Use With (Equipment) | 20 mm Headspace Septa - MicroLiter | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction