Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences MicroLiter 9 mm ILT LC-Clean™ Screw Thread Autosampler Vial Caps with PTFE/Silicone Septa
SDP

Catalog No. p-7237564 Shop All DWK Life Sciences Products
Click to view available options
Color:
Blue
Gray

Available in multiple colors and package sizes.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polypropylene
Pre-slit Yes
Septum PTFE, Silicone
No. per Pack 100
Product Line MicroLiter
Type Screw Cap
Closure Size 9 mm
For Use With (Equipment) 9 mm Autosampler Vials - MicroLiter
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.