Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Microliter Sample Syringes - Gas Phase
GREENER_CHOICESDP

Catalog No. 31502307 Shop All Kemtech Products
Change view
Click to view available options
Volume (Metric):
0.5 μL
1 μL
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1000 μL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
31-502-307 100 μL
31-502-301 0.5 μL
31-502-302 1 μL
31-502-303 5 μL
31-502-304 10 μL
31-502-305 25 μL
31-502-306 50 μL
31-502-308 250 μL
31-502-309 500 μL
31-502-310 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 31-502-307 Supplier Synthware™ Supplier No. S371007
Add to Cart
Add to Cart

Borosilicate syringes with a beveled tip, for precise sample delivery in volumes ranging from 0.5 μL to 1 mL.

  • Borosilicate glass syringes with a beveled tip
  • For precise sample delivery
  • Gas phase

Specifications

Material Borosilicate Glass (3.3)
Color Clear
For Use With (Application) Precise Sampling
Latex Free Yes
Product Type Syringe
Disposable Reusable
Volume (Metric) 100 μL
Needle Point Style Bevel
For Use With (Equipment) Syringe Needles
Connection Type Slip Tip
Graduated Yes
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.