Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks for Primera Printers
Catalog No. 22038028 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-028 Peach
22-038-023 White
22-038-024 Pink
22-038-025 Green
22-038-026 Yellow
22-038-027 Blue
22-038-029 Tan
22-038-030 Gray
22-038-031 Lilac
22-038-032 Orange
22-038-033 Aqua
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-038-028 Supplier Simport™ Scientific Supplier No. M4077T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Secure specimens during processing with Micromesh™ Biopsy Processing/Embedding Cassettes in Quickload™ Stacks for Primera Printers. No need for biopsy pads thanks to the 1676 square 0.36 mm openings of the mesh.
  • In Quickload™ stacks, for Primera printers
  • 45° Anterior writing area
  • 1676 square opening (0.36 mm)
  • No biopsy pads needed
  • 1 large 27 mm compartment
  • Recessed cover
  • Suitable for automated printers
  • Made of acetal
  • Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Quantity 2000/Pk.
Color Peach
Material Acetal