Learn More
Simport™ Scientific Micromesh™ Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeves
Catalog No. 23045367 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
23-045-367 Pink
23-045-363 Lilac
23-045-364 Orange
23-045-365 Aqua
23-045-366 White
23-045-368 Green
23-045-369 Yellow
23-045-370 Blue
23-045-371 Peach
23-045-373 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 23-045-367 Supplier Simport™ Scientific Supplier No. M5073SL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Micromesh™ Biopsy Processing/Embedding Cassettes in QuickLoad™ Sleeves are molded from a special high density polymer to keep specimens safely submerged in liquid.
  • Totally resistant to the chemical action of histological solvents.
  • Lid can be opened and closed as often as necessary and always re-locks securely without danger of specimen loss.
  • Offers 1,676 square openings (0.38 mm) allowing for a greatly improved fluid exchange and drainage without having to use biopsy pads.
  • Large anterior slots in both cassette and cover ensure that the Micromesh™ biopsy cassette will sink rapidly.
  • Large square compartment measuring 27 mm is ideal even for needle biopsies.
  • Cover does not protrude above the cassette, an excellent space-saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
  • Secure locking device safely holds the specimen during processing.
  • Convenient Quickload™ sleeves are suitable for automated printers.
Specifications
Quantity 75 sleeves/Cs.
Writing Face Angle 45°
Color Pink
Material Acetal
Product Type Cassette