Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Micronova™ Cleanroom Barrier Tapes

Shop All Micronova Manufacturing Products


Click to view available options
Color:
Red
Yellow
Display Text:
Caution
Do Not Enter
Description
The Micronova Cleanroom Barrier Tapes help prevent accidents by increasing visibility of hazards and can be tied, stapled or nailed in place. Roll text is bold, 2-inch high lettering in English and Spanish; available in either red - “Do Not Enter”, or yellow - “Caution”.
- Barrier tapes for increased visibility
- 1000 ft. of 3 mil non-adhesive polyethylene film
- For blocking off restricted areas in the cleanroom
- Bold black text is in English and Spanish
- Available in either red or yellow film
- Red tape reads: DANGER DO NOT ENTER/PELIGRO NO ENTRAR
- Yellow tape reads: CAUTION/CUIDADO
- Wound on a 3-inch plastic core
Specifications
Specifications
Autoclavable | Not Autoclavable |
Adhesive Type | Non-adhesive |
Color | Yellow |
Material | Polyethylene |
Product Type | Cleanroom Barricade Tape |
Sterility | Non-sterile |
Display Text | Caution |
For Use With (Application) | Recommendation to Use Caution for Safety |
Temperature Range (English) | 60°F to 80°F |
Temperature Range (Metric) | 16°C to 27°C |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction